missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Biotin-X, SE (6-((Biotinoyl)Amino)Hexanoic Acid, Succinimidyl Ester (Biotinamidocaproate, N-Hydroxysuccinimidyl Ester))

Product Code. 11574157
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 11574157

Brand: Invitrogen™ B1582

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Biotin-X, SE (6-((Biotinoyl)Amino)Hexanoic Acid, Succinimidyl Ester (Biotinamidocaproate, N-Hydroxysuccinimidyl Ester))

The amine-reactive biotin-X, SE can be used to attach this important hapten to biomolecules of interest for subsequent detection with streptavidin, avidin or NeutrAvidin™ biotin-binding protein.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 mg
Product Type Biotin-XX SE
Content And Storage Store in freezer (-5 to -30°C).
Chemical Reactivity Amine
Shipping Condition Room Temperature
Reactive Moiety Active Ester, Succinimidyl Ester
Label or Dye Biotin-X
Label Type Biotin and Other Haptens

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.