missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Biotin XX Tyramide SuperBoost™ Kit, Streptavidin

Product Code. 15585055
Change view
Click to view available options
Quantity:
150 Slides
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15585055 150 Slides 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15585055 Supplier Invitrogen™ Supplier No. B40931
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

SuperBoost™ tyramide signal amplification is the most sensitive method for detection of low abundant targets in multiplexable fluorescent immunocytochemistry (ICC), immunohistochemistry (IHC ), and in situ hybridization (ISH).

SuperBoost™ tyramide signal amplification is the most sensitive method for detection of low abundant targets in multiplexable fluorescent immunocytochemistry (ICC), immunohistochemistry (IHC ), and in situ hybridization (ISH). SuperBoost kits combine the brightness of AlexaFluor™ dyes with the superior signal amplification of a poly-HRP-mediated tyramide labeling reaction to produce a sensitivity 10-200 times greater than standard methods. For standout research, SuperBoost kits sharpen your results for clear visibility into critical areas that standard imaging methods fail to reveal.

SuperBoost kits are simple to use and easily adapted to standard ICC, IHC, or FISH experimental protocols, using any cell or tissue type. Cells labeled using a SuperBoost kit can be imaged using any type of microscope, producing high-resolution multiplex images. This particular kit features biotin XX tyramide with HRP-conjugated streptavidin. For detection and additional signal amplification, conjugated streptavidin is used to detect specifically deposited biotin XX .

Features of the SuperBoost kits include:
• Superior sensitivity for detection of low-level or hard-to-detect targets by fluorescent imaging
• Simple protocol and detection using standard filters
• Suitable for high-resolution multiplex images—co-label with DAPI, secondary antibodies, and other SuperBoost kits
• Requires 10-100 times less primary antibody then standard ICC/IHC/ISH experiments

SuperBoost kits are based on the tyramide signal amplification system, which uses the catalytic activity of horseradish peroxidase (HRP) to generate high density labeling of a target protein or nucleic acid sequence in situ. A typical ICC/IHC/ISH experiment using a SuperBoost kit requires 10-100 times less primary antibody then standard ICC/IHC/ISH experiments. SuperBoost kits offer superior specific signal intensity over background, so the protocol is easily optimized to detect specific signal in samples where high endogenous autofluorescence is observed.

TRUSTED_SUSTAINABILITY

Specifications

Conjugate Biotin-XX
Quantity 150 Slides
Content And Storage 1 kit sufficient for 150 microscope slides (18 x 18 mm), containing: Blocking buffer (1 X), 22.5 mL
  • HRP-conjugated streptavidin (1 X), 22.5 mL
  • Biotin-XX tyramide reagent
  • Hydrogen peroxide (stabilized 3% solution
  • Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
    Product Line SuperBoost
    Product Type Tyramide Kit

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.