missing translation for 'onlineSavingsMsg'
Learn More

Biotinyl-6-aminoquinoline, TRC

Product Code. 30240592
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30240592 100 mg 100mg
30240909 250 mg 250mg
30240637 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30240592 Supplier Toronto Research Chemicals Supplier No. TRCB016400100MG
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91853-89-3
Molecular Formula C19H22N4O2S
Molecular Weight (g/mol) 370.47
Synonym (3aS,4S,6aR)-Hexahydro-2-oxo-N-6-quinolinyl-1H-thieno[3,4-d]imidazole-4-pentanamide;
IUPAC Name 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]-N-quinolin-6-ylpentanamide
SMILES O=C(CCCC[C@H]1SC[C@H]2NC(=O)N[C@@H]12)Nc3ccc4ncccc4c3

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C19H22N4O2S/c24-17(21-13-7-8-14-12(10-13)4-3-9-20-14)6-2-1-5-16-18-15(11-26-16)22-19(25)23-18/h3-4,7-10,15-16,18H,1-2,5-6,11H2,(H,21,24)(H2,22,23,25)/t15-,16-,18-/m0/s1
Formula Weight 370.1463
Chemical Name or Material Biotinyl-6-aminoquinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.