missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Biotinyl Tyramide

Product Code. 18757506
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18757506 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18757506 Supplier R&D Systems™ Supplier No. 6241/25
  • ONLINE EXCLUSIVE PRICE
    128.91 €
    115.50 € / 25mg
    Save 13.41 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Reagent widely used for signal amplification in IHC and FISH

Biotinyl Tyramide is a reagent widely used for signal amplification in IHC and fluorescent in situ hybridization (ISH). HRP catalyzes localized deposition of multiple biotinyl tyramide molecules (catalyzed reporter deposition, CARD), which can be detected using a labeled streptavidin conjugate. The localization of deposition around HRP leads to good resolution. Suitable for either fluorescence or chromogenic detection.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (3 AS,4 S,6 aR)-Hexahydro-N-[2-(4-hydroxyphenyl)ethyl]-2-oxo-1 H-thieno[3,4-d]imidazole-4-pentanamide
CAS 41994-02-9
Synonym (3aS,4S,6aR)-Hexahydro-N-[2-(4-hydroxyphenyl)ethyl]-2-oxo-1H-thieno[3,4-d]imidazole-4-pentanamide
Quantity 25 mg
Product Line Tocris
Target Tyramide Signaling Amplification/TSA
Molecular Formula C18H25N3O3S
Formula Weight 363.47
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.