missing translation for 'onlineSavingsMsg'
Learn More

Biotinyl Tyramide, TRC

Product Code. 30133131
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133131 100 mg 100mg
30003617 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133131 Supplier Toronto Research Chemicals Supplier No. TRCB397770100MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41994-02-9
Molecular Formula C18 H25 N3 O3 S
Molecular Weight (g/mol) 363.47
Synonym [3aS-(3aα,4β,6aα)]-Hexahydro-N-[2-(4-hydroxyphenyl)ethyl]-2-oxo-, 1H-thieno[3,4-d]imidazole-5-pentanamide;
IUPAC Name 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]-N-[2-(4-hydroxyphenyl)ethyl]pentanamide
SMILES Oc1ccc(CCNC(=O)CCCC[C@@H]2SC[C@@H]3NC(=O)N[C@H]23)cc1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C18H25N3O3S/c22-13-7-5-12(6-8-13)9-10-19-16(23)4-2-1-3-15-17-14(11-25-15)20-18(24)21-17/h5-8,14-15,17,22H,1-4,9-11H2,(H,19,23)(H2,20,21,24)/t14-,15-,17-/m0/s1
Formula Weight 363.1617
Percent Purity >95%
Chemical Name or Material Biotinyl Tyramide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.