missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Biotinylated Human CD3 epsilon & CD3 delta Heterodimer Protein, Fc,His,Avitag™ & Fc,Flag,Avitag™

Product Code. 17176211
Click to view available options
Quantity:
200 μg
25 μg
Unit Size:
200µg
25µg

Product Code. 17176211

Brand: ACROBiosystems CDDH82F625ug

Please to purchase this item. Need a web account? Register with us today!

MALS verified

T-cell surface glycoprotein CD3 delta & CD3 epsilon chain, also known as CD3D & CD3E or CD3D&CD3E respectively, are single-pass type I membrane proteins. CD3D, together with CD3- epsilon(CD3E) , CD3-gamma and CD3-zeta, and the T-cell receptor alpha/beta and gamma/delta heterodimers, forms the T cell receptor-CD3 complex. T cell receptor-CD3 complex plays an important role in coupling antigen recognition to several intracellular signal-transduction pathways.

Biotinylated Human CD3E&CD3D Heterodimer Protein, Fc,His,Avitag&Fc,Flag,Avitag is produced by co-expression of CD3E and CD3D, has a calculated MW of 45.1kDa (CD3E) & 42.6 kDa (CD3D). Subunit CD3E is fused with a human IgG1 Fc fragment at the C-terminus, f

Specifications

Accession Number NP_000724.1 (CD3E) and NP_000723.1 (CD3D)
Molecular Weight (g/mol) 45.1 kDa (CD3E) and 42.6 kDa (CD3D)
Quantity 25 μg
Source MABSol™ Biotinylated Human CD3E&CD3D; Heterodimer Protein, Fc,His,Avitag&Fc;,Flag,Avitag (CDD-H82F6) is expressed from human HEK293 cells. It contains AA Asp 23 - Asp 126 (CD3E) and Phe 22 - Ala 105 (CD3D) (Accession # P07766-1 (CD3E) and P04234-1 (CD3D)). It is the biotinylated form of Human CD3 epsilon and CD3 delta Protein.
Immunogen Asp 23 - Asp 126 (CD3E) and Phe 22 - Ala 105 (CD3D)
Storage Requirements -20°C
Gene Alias CD3E and CD3D,CD3 delta and CD3 epsilon,CD3 delta / epsilon
Common Name CD3E and CD3D
Conjugate Precision Avi
Species Human
Protein Tag Fc Tag and His Tag and Flag Tag and Avi Tag
Buffer PBS, pH7.4
Endotoxin Level Less than 1.0 EU / μg by the LAL method.
Expression System HEK293
Form Powder
Protein Subtype MABSol™ Biotin Labeled Proteins
Purity or Quality Grade >90% as determined by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.