Learn More
ACROBiosystems Biotinylated Human IL-4 R alpha / CD124 Protein, Avitag™,His Tag
Description
IL-4 R alpha is also known as IL4R, CD124, IL-4RA.This gene encodes the alpha chain of the interleukin-4 receptor, a type I transmembrane protein that can bind interleukin 4 and interleukin 13 to regulate IgE antibody production in B cells. Allelic variations in this gene have been associated with atopy, a condition that can manifest itself as allergic rhinitis, sinusitis, asthma, or eczema. The binding of IL-4 or IL-13 to the IL-4 receptor on the surface of macrophages results in the alternative activation of those macrophages. Interleukin-4 receptor has been shown to interact with SHC1.
Specifications
Specifications
| Accession Number | NP_000409.1 |
| Molecular Weight (g/mol) | 26.9 kDa |
| Quantity | 200 μg |
| Source | Biotinylated Human IL-4 R alpha, Avitag,His Tag (ILR-H82E9) is expressed from human 293 cells (HEK293). It contains AA Met 26 - His 232 (Accession # NP_000409.1). |
| Immunogen | Met 26 - His 232 |
| Storage Requirements | -20°C |
| Gene Alias | IL4R,CD124,IL4RA |
| Common Name | IL-4 R alpha |
| Conjugate | Precision Avi |
| Species | Human |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.