missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Biotinylated Human IL-4 R alpha / CD124 Protein, Avitag™,His Tag

Product Code. p-7215521
Change view
Click to view available options
Quantity:
200 μg
25 μg
Unit Size:
200µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15847127 200 μg 200µg
15857127 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15847127 Supplier ACROBiosystems Supplier No. ILRH82E9200UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Biotinylated Protein

IL-4 R alpha is also known as IL4R, CD124, IL-4RA.This gene encodes the alpha chain of the interleukin-4 receptor, a type I transmembrane protein that can bind interleukin 4 and interleukin 13 to regulate IgE antibody production in B cells. Allelic variations in this gene have been associated with atopy, a condition that can manifest itself as allergic rhinitis, sinusitis, asthma, or eczema. The binding of IL-4 or IL-13 to the IL-4 receptor on the surface of macrophages results in the alternative activation of those macrophages. Interleukin-4 receptor has been shown to interact with SHC1.

This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 26.9 kDa. The protein migrates as 40-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.

Specifications

Accession Number NP_000409.1
Molecular Weight (g/mol) 26.9 kDa
Quantity 200 μg
Source Biotinylated Human IL-4 R alpha, Avitag,His Tag (ILR-H82E9) is expressed from human 293 cells (HEK293). It contains AA Met 26 - His 232 (Accession # NP_000409.1).
Immunogen Met 26 - His 232
Storage Requirements -20°C
Gene Alias IL4R,CD124,IL4RA
Common Name IL-4 R alpha
Conjugate Precision Avi
Species Human
Protein Tag His Tag and Avi Tag
Buffer PBS, pH7.4
Endotoxin Level Less than 1.0 EU / μg by the LAL method.
Expression System HEK293
Form Powder
Protein Subtype MABSol™ Biotin Labeled Proteins
Purity or Quality Grade >95% as determined by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.