missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Specific for Matrix Metalloproteinase-2 (MMP-2)
- Either the endogenous (naturally occurring) or total (following activation using APMA) activity levels in samples can be detected
- Range and sensitivity equivalent to ELISA
- Non-radioactive coated microplate format for ease of use
- Measures active MMPs from a range of species, including human, mouse, rat, rabbit and porcine
- Applicable for complex biological samples such as serum, plasma, synovial fluids, tissue extracts, cell culture supernatants
- Flexible protocol to cover speed and sensitivity requirements
Specifications
Specifications
| Antigen | MMP-2 |
| Assay Range | 0.75 ng/ml/ (0.19 ng/ml possible with longer incubation)-12 ng/ml (3 ng/ml possibile with longer incubation) |
| Description | Matrix Metalloproteinase-2, Biotrak Assay |
| Molecular Weight (g/mol) | 66000 |
| Quantity | 96-wells |
| Sample Volume | 100 μL |
| Sensitivity | 0.5 ng/mL (short incubation), 190 pg/mL (longer incubation) |
| Content And Storage | -15°C to -30°C |
| For Use With (Application) | ELISA |
| Specificity | The assay recognises the pro and active forms of MMP-2. It does not cross-react with other MMPs or TIMPs. Cross-reactivity has also been shown with rabbit, porcine, rat and mouse samples. Both pro and active MMP-2/TIMP-2 complexes as well as active MMP-2/ |
Safety and Handling
Signal Word
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Acute toxicity Category 4
- Acute toxicity Category 2
- Acute toxicity Category 1
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H412-Harmful to aquatic life with long lasting effects.
- H319-Causes serious eye irritation.
- H315-Causes skin irritation.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H300-Fatal if swallowed.
- H310-Fatal in contact with skin.
- H330-Fatal if inhaled.
- H373-May cause damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P271-Use only outdoors or in a well-ventilated area.
- P270-Do not eat, drink or smoke when using this product.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P262-Do not get in eyes, on skin, or on clothing.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302-if on skin: .
- P405-Store locked up.
- P501-Dispose of contents/container to .
- MIXTURE LIST-Contains : 2-amino-2-(hydroxymethyl)propane-1, 3-diol hydrochloride, zinc chloride, trometamol, S-2444 substrat
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?