missing translation for 'onlineSavingsMsg'
Learn More

Biochrom Biowave 3

Product Code. 16610864
Change view
Click to view available options
Includes:
Spec, Bluetooth Connectivity
Spec, Bluetooth, Printer
Spec, Built-in Serial Printer
Spectrophotometer
Interface:
Large Disp,Button UI,Bluetooth
Large Display, Button UI
Printer Included:
No
Yes
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Interface Printer Included unitSize
16610864 Spectrophotometer Large Display, Button UI No Each
17293054 Spec, Built-in Serial Printer Large Display, Button UI Yes Each
17203064 Spec, Bluetooth Connectivity Large Disp,Button UI,Bluetooth No Each
17213064 Spec, Bluetooth, Printer Large Disp,Button UI,Bluetooth Yes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16610864 Supplier Biochrom Supplier No. 80300732
Discount Lab Supplies
Use promo code 28170
View all promotions
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

The Biowave 3 offers all the capabilities of a research grade spectrophotometer with a robust design and a reasonable cost make it great for students.

  • UV-Vis Spectrophotometer with a 5 nm bandwidth
  • Features a USB port for data collection
  • Pre-programmed life science methodologies, including nucleic acid quantification (DNA, RNA and oligonucleotides), protein assays, and for cell culture density (OD600) measurements
  • Nucleic acid scans for purity checking
  • Low volume capability for use with microvolume cuvettes
  • Novel optics with a Xenon source for long lamp lifetime and low running costs
  • Choose to include a built-in serial printer, Bluetooth connectivity, or both
  • Lightweight and stackable for storage
  • Spectrophotometer has a 2 year warranty and the Xenon flash lamp has a 3 year warranty
  • IQ/OQ available on request

Specifications

Beam Center Height 15 mm
Depth (English) 15.3 in.
Depth (Metric) 390 mm
Detector Type Twin CMOS Array
Display Graphic LCD
Languages English, French, German, Spanish, Italian, Chinese, Japanese
Light Source Xenon Flash
Product Line Biowave 3
Spectral Bandwidth 5 nm
Type UV-Vis Spectrophotometer
Voltage 100/115/220/230 V
Wavelength Range 190 to 1100 nm
For Use With (Application) UV/Vis Spectrophotometry
For Use With (Equipment) Microvolume Cuvettes
Height (English) 3.9 in.
Height (Metric) 100 mm
Includes Spectrophotometer
Interface Large Display, Button UI
Monochromator Flat Grating
Noise ±0.005 peak to peak ±0.002 RMS
Photometric Linearity ±1.3% or ±0.008 A (Whichever is Greater at 546 nm)
Photometric Range -0.3 to 2.5 A, 0.3 to 199 %T
Photometric Repeatability ±0.002 A to 0.5 A (at 546 nm)
Printer Included No
Shape Round
Stray Light >0.5 %T 340 nm
Wavelength Accuracy ±2 nm
Wavelength Repeatability ±1 nm
Weight (English) 6.6 lb.
Weight (Metric) 3 kg
Width (English) 10 in.
Width (Metric) 260 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.