missing translation for 'onlineSavingsMsg'
Learn More

Biphenyl-4,4'-diboronic acid bis(pinacol) ester, 95%

Product Code. 15499465
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15499465 1 g 1g
15409475 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15499465 Supplier Thermo Scientific Alfa Aesar Supplier No. H51956.03
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 207611-87-8 | C24H32B2O4 | 406.14 g/mol

Biphenyl-4,4'-diboronic acid bis(pinacol) ester is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Biphenyl-4,4′-diboronic acid bis(pinacol) ester is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207611-87-8
Molecular Formula C24H32B2O4
Molecular Weight (g/mol) 406.14
MDL Number MFCD08669545
InChI Key INNVTCAYPFZCAL-UHFFFAOYSA-N
Synonym 4,4'-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1,1'-biphenyl, 4,4'-biphenyldiboronic acid dipinacol ester, 4,4'-biphenyldiboronic acid pinacol ester, 4,4'-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl biphenyl, biphenyl-4,4'-diboronic acid bis pinacol ester, 4,4'-biphenyldiboronic acid bis pinacol ester, 4,4,5,5-tetramethyl-2-4'-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1,1'-biphenyl-4-yl-1,3,2-dioxaborolane, amtb257, 4,4'-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl biphenyl
PubChem CID 10525359
IUPAC Name 4,4,5,5-tetramethyl-2-[4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]phenyl]-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Specifications

Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 406.14
Percent Purity 95%
Chemical Name or Material Biphenyl-4,4'-diboronic acid bis(pinacol) ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.