missing translation for 'onlineSavingsMsg'
Learn More

Biphenyl-4-boronic acid pinacol ester, 97%

Product Code. 15436368
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15436368 1 g 1g
15446368 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15436368 Supplier Thermo Scientific Alfa Aesar Supplier No. H59919.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 144432-80-4 | C18H21BO2 | 280.17 g/mol

It is an important raw material and intermediate used in organic synthesis and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis and pharmaceuticals.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144432-80-4
Molecular Formula C18H21BO2
Molecular Weight (g/mol) 280.17
MDL Number MFCD07368291
InChI Key REDKQKNJWVIPIO-UHFFFAOYSA-N
Synonym 4-biphenylboronic acid pinacol ester, 2-1,1'-biphenyl-4-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-4-biphenylyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-biphenylboronic acid, pinacol ester, 4,4,5,5-tetramethyl-2-4-phenylphenyl-1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 2-1,1'-biphenyl-4-yl-4,4,5,5-tetramethyl, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl biphenyl, 2-biphenyl-4-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl diphenyl
PubChem CID 11989651
IUPAC Name 4,4,5,5-tetramethyl-2-(4-phenylphenyl)-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 280.17
Percent Purity 97%
Chemical Name or Material Biphenyl-4-boronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.