missing translation for 'onlineSavingsMsg'
Learn More

Biphenyl-4-carbonyl chloride, 98%

Product Code. 11491698
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491698 10 g 10g
11401708 50 g 50g
11411708 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11491698 Supplier Thermo Scientific Alfa Aesar Supplier No. A13956.09
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14002-51-8 | C13H9ClO | 216.66 g/mol

Biphenyl-4-carbonyl chloride was used in the preparation of a novel thiourea compound, N-(6-methyl pyridin-2-yl-carbamothioyl)biphenyl-4-carboxamide. It was also used in the preparation of 5-CF3-oxazole analog, 2-{4-[2-(2-biphenyl-4-yl-5-trifluoromethyl-oxazol-4-yl)-ethoxy]-phenoxy}-2-methyl-propionic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Biphenyl-4-carbonyl chloride was used in the preparation of a novel thiourea compound, N-(6-methyl pyridin-2-yl-carbamothioyl)biphenyl-4-carboxamide. It was also used in the preparation of 5-CF3-oxazole analog, 2-{4-[2-(2-biphenyl-4-yl-5-trifluoromethyl-oxazol-4-yl)-ethoxy]-phenoxy}-2-methyl-propionic acid.

Solubility
Hydrolysis in water.

Notes
Moisture sensitive. Incompatible with oxidizing agent, bases, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14002-51-8
Molecular Formula C13H9ClO
Molecular Weight (g/mol) 216.66
MDL Number MFCD00000692
InChI Key JPVUWCPKMYXOKW-UHFFFAOYSA-N
Synonym 4-biphenylcarbonyl chloride, 1,1'-biphenyl-4-carbonyl chloride, biphenyl-4-carbonyl chloride, p-phenylbenzoyl chloride, 4-biphenylcarbonylchloride, 4-phenyl benzoyl chloride, 4-biphenyl carbonyl chloride, 1,1'-biphenyl-4carbonyl chloride
PubChem CID 84151
IUPAC Name 4-phenylbenzoyl chloride
SMILES ClC(=O)C1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point 110°C to 115°C
Boiling Point 160°C (2 mmHg)
Quantity 10 g
UN Number UN3261
Beilstein 472842
Sensitivity Moisture sensitive
Solubility Information Hydrolysis in water.
Formula Weight 216.67
Percent Purity 98%
Chemical Name or Material Biphenyl-4-carbonyl chloride
compliance-icons
Product Identifier
  • Biphenyl-4-carbonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: [1,1-Biphenyl]-4-carbonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.