missing translation for 'onlineSavingsMsg'
Learn More

Biphenylene, 99+%, Thermo Scientific Chemicals

Product Code. 10712491 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 mg
250 mg
Packaging:
Glass bottle
Unit Size:
250mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10712491 250 mg Glass bottle 250mg
10381474 50 mg Glass bottle 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10712491 Supplier Thermo Scientific Acros Supplier No. 279742500
 Restricted Item
Estimated Shipment: 23-11-2026
Aggiungi al carrello
Aggiungi al carrello
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 259-79-0 | Molecular Formula: C12H8 | Molecular Weight: 152.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 259-79-0
Molecular Formula C12H8
Molecular Weight (g/mol) 152.2
MDL Number MFCD00001110
InChI Key IFVTZJHWGZSXFD-UHFFFAOYSA-N
Synonym diphenylene, cyclobutadibenzene, dibenzocyclobutadiene, 1,1'-biphenylene, unii-0z64i7d5m2, 8ci 9ci, biphenylen, biphenylene, biphenylene, 99+%, pubchem9106
PubChem CID 9214
ChEBI CHEBI:33079
IUPAC Name biphenylene
SMILES C1=CC=C2C(=C1)C3=CC=CC=C23

Specifica

CAS Min % 99.0
CAS Max % 100.0
Melting Point 109°C to 112°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 250 mg
Beilstein 05, 298
Solubility Information Solubility in water: 9.60mg/l (calc). Other solubilities: organic solvents general
Formula Weight 152.2
Percent Purity 99+%
Physical Form Needles
Chemical Name or Material Biphenylene, 99+%
Vedi altri risultati Mostra meno risultati
compliance-icons
Identificativo prodotto
  • Biphenylene
Categoria di pericolo
  • LONG-TERM AQUATIC HAZARD Chronic 2
Indicazioni di pericolo
  • H411-Toxic to aquatic life with long lasting effects.
Consigli di prudenza
  • P273-Avoid release to the environment.

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.