missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ BIRB 796
GREENER_CHOICE

Product Code. 18757996
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
50 mg
missing translation for 'unitSize'
10mg
50mg
This item is not returnable. View return policy

Product Code. 18757996

missing translation for 'mfr': Tocris Bioscience™ 5989/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High affinity and selective p38 kinase inhibitor

BIRB 796 is a high affinity and selective p38 kinase inhibitor (Kd = 50-100 pM). Exhibits no significant inhibition on a panel of related kinases. Inhibits LPS-induced TNFα production in human PBMCs and whole blood (IC50 values are 21 and 960 nM, respectively). Also inhibits JNK2α2 and c-Raf-1 (IC50 values are 98 nM and 1.4 μM, respectively). Cell permeable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[3-(1,1-Dimethylethyl)-1-(4-methylphenyl)-1H-pyrazol-5-yl]-N′-[4-[2-(4-morpholinyl)ethoxy]-1-naphthalenyl]urea
CAS 285983-48-4
Synonym Doramapimod
Quantity 10 mg
Target p38 Inhibitors
Molecular Formula C31H37N5O3
Formula Weight 527.66
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.