Learn More
Birkenstock™ Birkenstock™ Bottle BOD
Product Code. 12307218
Encompass
Change view
Click to view available options
Volume (Metric):
500 mL
Unit Size:
Each
Product Code. Volume (Metric) Unit Size
12307218 500 mL Each
1 options
Product Code. 12307218 Supplier Birkenstock™ Supplier No. GS748/B
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
With interchangable glass peg-stopper with cone point, adjusted to leave a 'well' for a water seal at the bottle opening.
Specifications
Closure Type Glass peg-stopper with cone point
Volume (Metric) 500 mL
For Use With (Application) BOD