missing translation for 'onlineSavingsMsg'
Learn More

Bis[1,2-bis(diphenylphosphino)ethane]palladium(0)

Product Code. 11343628
Change view
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11343628 1 g 1g
11333628 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11343628 Supplier Thermo Scientific Alfa Aesar Supplier No. 039233.03
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 31277-98-2 | C52H48P4Pd | 903.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31277-98-2
Molecular Formula C52H48P4Pd
Molecular Weight (g/mol) 903.27
MDL Number MFCD00009880
InChI Key FAFGMAGIYHHRKN-UHFFFAOYSA-N
Synonym bis 1,2-bis diphenylphosphino ethane palladium 0, pd dppe 2, pd diphos 2, bis diphos palladium 0, bis 1,2-bis diphenylphosphino ethane palladium, bis diphos palladium, bis 1,2-bis diphenylphosphino ethane-palladium 0, bis 1,2-bis diphenylphosphino ethane-palladium
PubChem CID 2724231
IUPAC Name 2-diphenylphosphanylethyl(diphenyl)phosphane;palladium
SMILES [Pd].C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 903.25
Physical Form Powder
Chemical Name or Material Bis[1,2-bis(diphenylphosphino)ethane]palladium(0)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.