missing translation for 'onlineSavingsMsg'
Learn More

Bis(1,5-cyclooctadiene)diiridium(I) Dichloride, TRC

Product Code. 30329323
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30329323 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30329323 Supplier Toronto Research Chemicals Supplier No. TRCB423265500MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bis(1,5-cyclooctadiene)diiridium(I) Dichloride
CAS 12112-67-3
Molecular Formula C16 H22 Cl2 Ir2
Quantity 500 mg
Synonym Chloro(1,5-cyclooctadiene)iridium Dimer, Chloro(cyclooctadiene)iridium Dimer, Cyclooctadieneiridium Chloride Dimer, Di-μ-Chlorobis(1,5-cyclooctadiene)diiridium, Di-μ-Chlorobis(π-1,5-cyclooctadiene)diiridium, Di-μ-chlorobis(eta4-1,5-cyclooctadiene)diiridium, Di-μ-chlorobis(eta4-1,5-cyclooctadiene)diiridium(I), Di-μ-chlorobis[(1,2,5,6-eta)-1,5-cyclooctadiene]diiridium, Di-μ-chlorobis[(1,5-cyclooctadiene)iridium], Di-μ2-chlorobis(1,5-cyclooctadiene)diiridium, Dichlorobis(1,5-cyclooctadiene)diiridium, Dichlorobis(cyclooctadiene)diiridium, Dichlorobis(eta4-1,5-cyclooctadiene)diiridium, NSC 326162, μ,μ-Dichlorobis(1,5-cyclooctadiene)diiridium, Bis(μ-chloro)bis(1,5-cyclooctadiene)diiridium, Bis[μ-chloro(eta4-1,5-cyclooctadiene)iridium], Bis(chloro(1,5-cyclooctadiene)iridium), Bis(chloro(cyclooctadiene)iridium), Bis(chloro(1,5-cyclooctadiene)iridium), Bis((μ-chloro)(1,5-cyclooctadiene)iridium), 5-Chlorobis(1,5-cyclooctadiene)diiridium, (eta4-1,5-Cyclooctadiene)iridium Chloride Dimer, (1,5-Cyclooctadiene)iridium Chloride Dimer, (1,5-Cyclooctadiene)chloroiridium Dimer, 1,5-Cyclooctadiene Iridium Complex
InChI Formula InChI=1S/2C8H10.2ClH.2Ir/c2*1-2-4-6-8-7-5-3-1;;;;/h2*1,8H,2,4-5,7H2;2*1H;;
SMILES C1CC2=CCCC(=C1)[Ir]23Cl[Ir]4(Cl3)C5=CCCC4=CCC5
IUPAC Name (1Z,5Z)-cycloocta-1,5-diene;iridium;dichloride
Molecular Weight (g/mol) 669.686
Formula Weight 670.036
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.