missing translation for 'onlineSavingsMsg'
Learn More

Bis(1,5-cyclooctadiene)nickel(0)

Product Code. 10548333 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10548333 5 g 5g
10126242 1 g 1g
11905831 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10548333 Supplier Thermo Scientific Acros Supplier No. 223970050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1295-35-8 | C16H24Ni | 275.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1295-35-8
Molecular Formula C16H24Ni
Molecular Weight (g/mol) 275.06
MDL Number MFCD00058902
InChI Key JRTIUDXYIUKIIE-KZUMESAESA-N
Synonym bis 1,5-cyclooctadiene nickel 0, bis 1,5-cyclooctadiene nickel, ni cod 2, ccris 5973, bis cyclooctadiene nickel 0, nickel, bis 1,5-cyclooctadiene, bis cyclooctadiene nickel, 1z,5z-cycloocta-1,5-diene; nickel, nickel biscod, pubchem16018
PubChem CID 6433264
SMILES [Ni].C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1

Specifications

Melting Point 60.0°C
Color Golden
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: insoluble.
IUPAC Name (1Z,5Z)-cycloocta-1,5-diene;nickel
Formula Weight 275.06
Physical Form Shiny Crystalline Powder
Chemical Name or Material Bis(1, 5-cyclooctadiene)nickel(0)
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Bis(1,5-cyclooctadiene)nickel(0)
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
  • Flammable solid Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H350-May cause cancer.
  • H228-Flammable solid.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.