missing translation for 'onlineSavingsMsg'
Learn More

Bis(2,2,2-trifluoroethyl)amine, 95%

Product Code. 11369345
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11369345 5 g 5g
11359345 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11369345 Supplier Thermo Scientific Alfa Aesar Supplier No. L16709.06
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 407-01-2 | C4H5F6N | 181.081 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 407-01-2
Molecular Formula C4H5F6N
Molecular Weight (g/mol) 181.081
MDL Number MFCD00042094
InChI Key GTJGHXLFPMOKCE-UHFFFAOYSA-N
PubChem CID 78999
IUPAC Name 2,2,2-trifluoro-N-(2,2,2-trifluoroethyl)ethanamine
SMILES C(C(F)(F)F)NCC(F)(F)F

Specifications

Density 1.1
Boiling Point 70°C to 71°C
Refractive Index 1.301
Quantity 5 g
UN Number UN2735
Beilstein 1811926
Synonym bis 2,2,2-trifluoroethyl amine, bis trifluoroethyl amine, 1,1,1,5,5,5-hexafluoro-3-azapentane, 2,2,2-trifluoro-n-2,2,2-trifluoroethyl ethanamine, ethanamine, 2,2,2-trifluoro-n-2,2,2-trifluoroethyl, bis trifluoroethyl-amin, acmc-1aha1, 1,1,15,5,5-hexafluoro-3-azapentane, ethanamine,2,2,2-trifluoro-n-2,2,2-trifluoroethyl, 2,2,2-tris fluoranyl-n-2,2,2-tris fluoranyl ethyl ethanamine
Formula Weight 181.08
Percent Purity 96%
Chemical Name or Material Bis(2,2,2-trifluoroethyl)amine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.