missing translation for 'onlineSavingsMsg'
Learn More

Bis(2,2,2-trifluoroethyl) phosphite, tech. 90%

Product Code. 15491858
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15491858 1 g 1g
15401868 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15491858 Supplier Thermo Scientific Alfa Aesar Supplier No. H55562.03
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 92466-70-1 | C4H4F6O3P+ | 245.037 g/mol

Bis(2,2,2-trifluoroethyl) phosphite was used in the synthesis of bis(2,2,2-trifluoroethyl phosphorochloridate. It was also employed as reagent for the synthesis of mono- and diesters of phosphorous acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(2,2,2-trifluoroethyl) phosphite was used in the synthesis of bis(2,2,2-trifluoroethyl phosphorochloridate. It was also employed as reagent for the synthesis of mono- and diesters of phosphorous acid.

Notes
Store at room temperature. Avoid strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92466-70-1
Molecular Formula C4H4F6O3P+
Molecular Weight (g/mol) 245.037
MDL Number MFCD00063314
InChI Key IMDCVAFSSZPRRM-UHFFFAOYSA-N
Synonym bis 2,2,2-trifluoroethyl phosphonate, phosphonic acid bis 2,2,2-trifluoroethyl ester, oxo-bis 2,2,2-trifluoroethoxy phosphanium, phosphonic acid, bis 2,2,2-trifluoroethyl ester, bis trifluoroethyl phosphonate, bis 2,2,2-trifluoroethyl phosphite, bis 2,2,2-trifluoroethyl hydrogen phosphite, oxobis 2,2,2-trifluoroethoxy-??-phosphanylium, bis 2,2,2-trifluoroethyl phosphite, technical grade
PubChem CID 6386648
IUPAC Name oxo-bis(2,2,2-trifluoroethoxy)phosphanium
SMILES C(C(F)(F)F)O[P+](=O)OCC(F)(F)F

Specifications

Density 1.545 g/mL
Boiling Point 43°C to 44°C (2 mmHg)
Flash Point 76°C (169°F)
Linear Formula (CF3CH2O)2P(O)H
Refractive Index 1.332
Quantity 1 g
Formula Weight 246.05
Percent Purity 90%
Grade Technical
Physical Form Liquid
Chemical Name or Material Bis(2,2,2-trifluoroethyl) phosphite
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.