missing translation for 'onlineSavingsMsg'
Learn More

Bis(2,2,6,6-tetramethyl-4-piperidyl)sebacate, TRC

Product Code. 30330751
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30330751 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30330751 Supplier Toronto Research Chemicals Supplier No. TRCB592180100MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sebacic Acid, Bis(2,2,6,6-tetramethyl-4-piperidinyl) Ester
CAS 52829-07-9
Molecular Formula C28 H52 N2 O4
Quantity 100 mg
Synonym Decanedioic acid, bis(2,2,6,6-tetramethyl-4-piperidinyl) ester (9CI), 770DF, ADK Stab LA 77, ADK Stab LA 77Y, Adeka LA 77, Adeka Stab LA 77Y, Adekastab LA 77, Antioxidant 770, BLS 1770, Bis(2,2,6,6-Tetramethyl-4-piperidone) sebacate, Bis(2,2,6,6-tetramethyl-4-piperidinyl) sebacate, Bis(2,2,6,6-tetramethyl-4-piperidyl) decanedioate, Bis(2,2,6,6-tetramethyl-4-piperidyl) sebacate, Bis(2,2,6,6-tetramethylpiperidin-4-yl) sebecate, Bis-(2,2,6,6-tetramethylpiperidin-4-yl) 1,10-decanedioate, Chisorb 770, Eversorb 90, GW 480, HA 10, HA 7OG, HALS 770, HS 770, Hillite 77, Hisorb 770, JF 90, Kemistab 77, LA 77, LA 77G, LA 77Y, LS 770, Light Stabilizer 770, Lowilite 77, MA 90, Mark LA 77, NF 90, Riasorb UV 770, SB-UVA 677, Sanol, Sanol 770, Sanol LS 107, Sanol LS 700, Sanol LS 770, Sanol LS 770P, Songlight 7700, Sumisorb 577, T 770, TIN 770, TN 770, Tinuvin 770, Tinuvin 770DF, Tinuvin 770DF1, Tinuvin 770LS, Tinuvin DF 770, UV 770, UV 770DF, Uvinul 4077, Uvinul 4077H, Viosorb 04, ZX 70, Sebacic acid, bis(2,2,6,6-tetramethyl-4-piperidinyl) ester, Sebacic Acid, Bis(2,2,6,6-tetramethyl-4-piperidinyl) Ester
InChI Formula InChI=1S/C28H52N2O4/c1-25(2)17-21(18-26(3,4)29-25)33-23(31)15-13-11-9-10-12-14-16-24(32)34-22-19-27(5,6)30-28(7,8)20-22/h21-22,29-30H,9-20H2,1-8H3
SMILES CC1(C)CC(CC(C)(C)N1)OC(=O)CCCCCCCCC(=O)OC2CC(C)(C)NC(C)(C)C2
IUPAC Name bis(2,2,6,6-tetramethylpiperidin-4-yl) decanedioate
Molecular Weight (g/mol) 480.72
Formula Weight 480.3927
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.