missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-benzamidophenyl) disulfide, 97%

Product Code. 11308625
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11308625

Brand: Thermo Scientific Alfa Aesar L12110.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 135-57-9 | C26H20N2O2S2 | 456.578 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135-57-9
Molecular Formula C26H20N2O2S2
Molecular Weight (g/mol) 456.578
MDL Number MFCD00043806
InChI Key ZHMIOPLMFZVSHY-UHFFFAOYSA-N
Synonym bis 2-benzamidophenyl disulfide, peptazin bafd, peptisant 1o, pepton 22, 2,2'-dibenzoylaminodiphenyl disulfide, bis o-benzamidophenyl disulfide, o,o'-dibenzamidodiphenyl disulfide, 2,2'-dibenzamidodiphenyl disulfide, n,n'-dithiodi-o-phenylenedibenzamide, 2',2'-dithiodibenzanilide
PubChem CID 67271
IUPAC Name N-[2-[(2-benzamidophenyl)disulfanyl]phenyl]benzamide
SMILES C1=CC=C(C=C1)C(=O)NC2=CC=CC=C2SSC3=CC=CC=C3NC(=O)C4=CC=CC=C4

Specifications

Melting Point 140°C to 143°C
Quantity 100 g
Beilstein 721443
Formula Weight 456.59
Percent Purity 97%
Chemical Name or Material Bis(2-benzamidophenyl) disulfide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.