missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-chloroethyl)amine hydrochloride, 98%

Product Code. 10438630 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10438630 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10438630 Supplier Thermo Scientific Acros Supplier No. 106321000
 Restricted Item
Estimated Shipment: 27-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 821-48-7 | C4H9Cl2N·HCl | 178.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 821-48-7
CAS Min % 97.5
CAS Max % 100.0
Melting Point 210°C to 215°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Molecular Formula C4H9Cl2N·HCl
Linear Formula (ClCH2CH2)2NH·HCl
MDL Number MFCD00012515
Quantity 100 g
Synonym bis 2-chloroethyl amine hydrochloride, 2,2'-dichlorodiethylamine hydrochloride, 2-chloro-n-2-chloroethyl ethanamine hydrochloride, nor-lost hydrochlorid, nor-hn2, ethanamine, 2-chloro-n-2-chloroethyl-, hydrochloride, n,n-bis 2-chloroethyl amine hydrochloride, di-2-chloroethyl amine hydrochloride, 2,2-dichlorodiethylamine hydrochloride, bis-2-chloroethyl amine hydrochloride
Solubility Information Solubility in water: soluble in water. Other solubilities: sparingly soluble in alcohols,insoluble in toluene and acetone
InChI Key YMDZDFSUDFLGMX-UHFFFAOYSA-N
SMILES C(CCl)NCCCl.Cl
IUPAC Name 2-chloro-N-(2-chloroethyl)ethanamine;hydrochloride
Molecular Weight (g/mol) 178.49
PubChem CID 522769
Formula Weight 178.49
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Bis(2-chloroethyl)amine hydrochloride, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Bis(2-chloroethyl)amine hydrochloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H341-Suspected of causing genetic defects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.