missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-chloroethyl)amine hydrochloride, 98%

Product Code. 11464464
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11464464 250 g 250g
11454464 50 g 50g
11444464 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11464464 Supplier Thermo Scientific Alfa Aesar Supplier No. A13118.30
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 821-48-7 | C4H10Cl3N | 178.481 g/mol

Bis(2-chloroethyl)amine hydrochloride is used as an intermediate in chemicals synthesis and building block for piperazine derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(2-chloroethyl)amine hydrochloride is used as an intermediate in chemicals synthesis and building block for piperazine derivatives.

Solubility
Partly miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 821-48-7
Molecular Formula C4H10Cl3N
Molecular Weight (g/mol) 178.481
MDL Number MFCD00012515
InChI Key YMDZDFSUDFLGMX-UHFFFAOYSA-N
Synonym bis 2-chloroethyl amine hydrochloride, 2,2'-dichlorodiethylamine hydrochloride, 2-chloro-n-2-chloroethyl ethanamine hydrochloride, nor-lost hydrochlorid, nor-hn2, ethanamine, 2-chloro-n-2-chloroethyl-, hydrochloride, n,n-bis 2-chloroethyl amine hydrochloride, di-2-chloroethyl amine hydrochloride, 2,2-dichlorodiethylamine hydrochloride, bis-2-chloroethyl amine hydrochloride
PubChem CID 522769
IUPAC Name 2-chloro-N-(2-chloroethyl)ethanamine;hydrochloride
SMILES C(CCl)NCCCl.Cl

Specifications

Melting Point 211°C to 215°C
Quantity 250 g
UN Number UN2923
Beilstein 3550356
Solubility Information Partly miscible in water.
Formula Weight 178.49
Percent Purity 98%
Chemical Name or Material Bis(2-chloroethyl)amine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.