missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-cyanoethyl)phenylphosphine, Thermo Scientific™

Product Code. 11393668
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11393668 1 g 1g
11383668 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11393668 Supplier Thermo Scientific Alfa Aesar Supplier No. 039268.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15909-92-9
Molecular Formula C12H13N2P
Molecular Weight (g/mol) 216.22
MDL Number MFCD00013827
InChI Key LIYGDNBFEBKBBZ-UHFFFAOYSA-N
Synonym bis 2-cyanoethyl phenylphosphine, 3,3'-phenylphosphinediyl dipropanenitrile, propanenitrile,3,3'-phenylphosphinidene bis, 3-2-cyanoethyl phenyl phosphanyl propanenitrile, acmc-20alo7, phenylbis 2-cyanoethyl phosphine, bis-beta-cyanoethyl-phenylphosphine, 3,3'-phenylphosphanediyl dipropanenitrile
PubChem CID 342156
IUPAC Name 3-[2-cyanoethyl(phenyl)phosphanyl]propanenitrile
SMILES N#CCCP(CCC#N)C1=CC=CC=C1

Specifications

Melting Point 72°C to 73°C
Boiling Point 176°C to 178°C (0.5 mmHg)
Quantity 1 g
UN Number UN3464
Formula Weight 216.22
Physical Form Crystalline
Chemical Name or Material Bis(2-cyanoethyl)phenylphosphine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.