missing translation for 'onlineSavingsMsg'
Learn More

Bis[2-(di-tert-butylphosphino)ethyl]amine, 10% w/w soln. in THF

Product Code. 15475796
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15475796 1 g 1g
15485796 5 g 5g
15495796 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15475796 Supplier Thermo Scientific Alfa Aesar Supplier No. H60493.03
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 944710-34-3 | C20H45NP2 | 361.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
This compound is involved in the transition metal-catalyzed addition of amines to acrylic acid derivatives.

Solubility
Insoluble in water.

Notes
Air Sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 944710-34-3
Molecular Formula C20H45NP2
Molecular Weight (g/mol) 361.54
MDL Number MFCD17014018
InChI Key MJYKYLNPIGDVEF-UHFFFAOYSA-N
Synonym bis 2-di-t-butylphosphino ethyl amine, bis 2-di-tert-butylphosphino ethyl amine, bis 2-di-tert-butylphosphanyl ethyl amine, bis 2-di-tert-butylphosphino ethyl amine w/w soln. in thf, bis 2-di-tert-butylphosphino ethyl amine solution, 10 wt. % in thf
PubChem CID 51346231
IUPAC Name 2-ditert-butylphosphanyl-N-(2-ditert-butylphosphanylethyl)ethanamine
SMILES CC(C)(C)P(CCNCCP(C(C)(C)C)C(C)(C)C)C(C)(C)C

Specifications

Density 0.911
Odor Unpleasant
Linear Formula ([(CH3)3C]2PCH2CH2)2NH
Refractive Index 1.418
Quantity 1 g
UN Number UN1993
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 361.53
Concentration or Composition (by Analyte or Components) 10% w/w soln. in THF
Chemical Name or Material Bis[2-(di-tert-butylphosphino)ethyl]amine
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Bis[2-(di-tert-butylphosphino)ethyl]amine, 10% w/w solution in THF
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Tetrahydrofuran, Bis[2-(di-tert-butylphosphino)ethyl]amine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.