missing translation for 'onlineSavingsMsg'
Learn More

Bis[2-(dicyclohexylphosphino)ethyl]amine, 97+%

Product Code. 15485606
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15485606 250 mg 250mg
15426748 1 g 1g
15436748 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15485606 Supplier Thermo Scientific Alfa Aesar Supplier No. H60269.MD
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 550373-32-5 | C28H53NP2 | 465.69 g/mol

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Air Sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 550373-32-5
Molecular Formula C28H53NP2
Molecular Weight (g/mol) 465.69
MDL Number MFCD17014019
InChI Key OQJIHPKBLHGKJN-UHFFFAOYSA-N
Synonym bis 2-dicyclohexylphosphino ethyl amine, bis 2-dicyclohexylphosphanyl ethyl amine, 2-dicyclohexylphosphanyl-n-2-dicyclohexylphosphanyl ethyl ethan-1-amine
PubChem CID 58194775
IUPAC Name 2-dicyclohexylphosphanyl-N-(2-dicyclohexylphosphanylethyl)ethanamine
SMILES C(CP(C1CCCCC1)C1CCCCC1)NCCP(C1CCCCC1)C1CCCCC1

Specifications

Melting Point 64°C to 69°C
Linear Formula [(C6H11)2PCH2CH2]2NH
Quantity 250 mg
UN Number UN3259
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 465.67
Percent Purity ≥97%
Chemical Name or Material Bis[2-(dicyclohexylphosphino)ethyl]amine
compliance-icons
Product Identifier
  • Bis[2-(dicyclohexylphosphino)ethyl]amine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Bis[2-(dicyclohexylphosphino)ethyl]amine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.