missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-diphenylphosphinoethyl)phenylphosphine, 97%

Product Code. 10003600 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10003600 1 g Glass bottle 1g
10301284 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10003600 Supplier Thermo Scientific Acros Supplier No. 316820010
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 23582-02-7 | C34H33P3 | 534.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23582-02-7
Molecular Formula C34H33P3
Molecular Weight (g/mol) 534.56
MDL Number MFCD00003048
InChI Key AXVOAMVQOCBPQT-UHFFFAOYSA-N
Synonym bis 2-diphenylphosphinoethyl phenylphosphine, phenylphosphinediyl bis ethane-2,1-diyl bis diphenylphosphine, bis 2-diphenylphosphino ethyl phenylphosphine, phosphine, bis 2-diphenylphosphino ethyl phenyl, phenylbis diphenylphosphinoethyl phosphine, pubchem6540, acmc-20alo6, chembl69711, bis 2-biphenylphosphinoethyl phenylphosphine
PubChem CID 90192
IUPAC Name bis(2-diphenylphosphanylethyl)-phenylphosphane
SMILES C(CP(C1=CC=CC=C1)C1=CC=CC=C1)P(CCP(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 121.0°C to 126.0°C
Color White
Elemental Analysis 74.1 to 78.7% (C)
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 534.56
Percent Purity 97%
Physical Form Chunks and/or Powder
Chemical Name or Material Bis(2-diphenylphosphinoethyl)phenylphosphine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.