missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-ethylhexyl)adipate, TRC

Product Code. 30005450
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
Unit Size:
100g
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005450 100 g 100g
30007849 25 g 25g
30006325 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005450 Supplier Toronto Research Chemicals Supplier No. TRCB433810100G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-23-1
Molecular Formula C22 H42 O4
Molecular Weight (g/mol) 370.57
Synonym Adipic acid, bis(2-ethylhexyl) ester (6CI,7CI,8CI), Hexanedioic acid, bis(2-ethylhexyl) ester (9CI), ADO, ADO (lubricating oil), Adimoll DO, Adipic acid diester with 2-ethylhexanol, Adipol 2EH, Arlamol DOA, Bis(2-ethylhexyl) adipate, Bisoflex DOA, Crodamol DOA, DOA, Dermol DOA, Di(2-ethylhexyl) adipate, Diacizer DOA, Diethylhexyl adipate, Dioctyl adipate, Dub Doa, Effomoll DA, Effomoll DOA, Ergoplast AdDO, Flexol A 26, Hatcol 2908, Jayflex DOA 2, K 3220, Kodaflex DOA, Lankroflex DOA, Monoplex DOA, NSC 56775, Octyl adipate, Plasthall DOA, Plastimoll DOA, Plastomoll DOA, Reomol DOA, SP 100, SP 100 (solvent), Sansocizer DOA, Sicol 250, Truflex DOA, USS 700, Vestinol OA, Vistone A 10, Wickenol 158, Witamol 320
IUPAC Name bis(2-ethylhexyl) hexanedioate
SMILES CCCCC(CC)COC(=O)CCCCC(=O)OCC(CC)CCCC

Specifications

Quantity 100 g
InChI Formula InChI=1S/C22H42O4/c1-5-9-13-19(7-3)17-25-21(23)15-11-12-16-22(24)26-18-20(8-4)14-10-6-2/h19-20H,5-18H2,1-4H3
Formula Weight 370.3083
Chemical Name or Material Adipic Acid Bis(2-ethylhexyl) Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.