missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(2-ethylhexyl) Azelate-d34, TRC
Specifications
Specifications
| Chemical Name or Material | Bis(2-ethylhexyl) Azelate-d34 |
| Molecular Formula | C25 D34 H14 O4 |
| Quantity | 5 mg |
| Synonym | Azelaic Acid Di(2-ethylhexyl) Ester-d34, Bis(2-ethylhexyl) Azelate-d34, Bis(2-ethylhexyl) Nonanedioate-d34, DOZ-d34, Di-2-ethylhexyl Azelate-d34, Dioctyl Azelate-d34, Dioctyl Aonanedioate-d34, Edenol 9058-d34, Emery 2958-d34, Emolien 2986-d34, Nonanedioic Acid Di(2-ethylhexyl) Ester-d34, Octyl Azelate-d34, Plastolein 9058-d34, Plastolein 9058DOZ-d34, Sansocizer DOZ-d34, Staflex DOX-d34, Truflex DOX-d34 |
| InChI Formula | InChI=1S/C25H48O4/c1-5-9-16-22(7-3)20-28-24(26)18-14-12-11-13-15-19-25(27)29-21-23(8-4)17-10-6-2/h22-23H,5-21H2,1-4H3/i1D3,2D3,3D3,4D3,5D2,6D2,7D2,8D2,9D2,10D2,16D2,17D2,20D2,21D2,22D,23D |
| SMILES | [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])(C([2H])([2H])OC(=O)CCCCCCCC(=O)OC([2H])([2H])C([2H])(C([2H])([2H])C([2H])([2H])[2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])C([2H])([2H])C([2H])([2H])[2H] |
| IUPAC Name | bis[1,1,2,3,3,4,4,5,5,6,6,6-dodecadeuterio-2-(1,1,2,2,2-pentadeuterioethyl)hexyl] nonanedioate |
| Molecular Weight (g/mol) | 446.856 |
| Formula Weight | 446.569 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?