missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-ethylhexyl) Phosphate, TRC

Product Code. 30136996
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136996 10 g 10g
30134187 5 g 5g
30003975 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136996 Supplier Toronto Research Chemicals Supplier No. TRCB43386510G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 298-07-7
Molecular Formula C16 H35 O4 P
Molecular Weight (g/mol) 322.42
Synonym 1-Hexanol, 2-ethyl-, hydrogen phosphate (7CI), 2-Ethyl-1-hexanol hydrogen phosphate, A 208, A 208 (phosphate), Baysolvex D 2EHPA, Bis(2-ethylhexyl) hydrogen phosphate, Bis(2-ethylhexyl) orthophosphate, Bis(2-ethylhexyl) phosphate, D 2EHPA, DEHAP, DP 8, DP 8R, Di(2-ethylhexyl) phosphate, Di(2-ethylhexyl)orthophosphoric acid, Di(2-ethylhexyl)phosphoric acid, Di-2-ethylhexyl hydrogen phosphate, Dioctyl hydrogen phosphate, Dioctyl orthophosphate, Dioctyl phosphate, HDEHP, HDEHPA, Hostarex PA 216, Hydrogen bis(2-ethylhexyl) phosphate, JB 58, LB 58, O,O-Bis(2-ethylhexyl)phosphoric acid, O,O-Di(2-ethylhexyl)phosphoric acid, Orthophosphoric acid 2-ethylhexyl alcohol diester, P 204, P 204 (acid), Phoslex A 208, Phosphoric acid di(2-ethylhexyl) ester, TOPS 99
IUPAC Name bis(2-ethylhexyl) hydrogen phosphate
SMILES CCCCC(CC)COP(=O)(O)OCC(CC)CCCC

Specifications

Quantity 10 g
InChI Formula InChI=1S/C16H35O4P/c1-5-9-11-15(7-3)13-19-21(17,18)20-14-16(8-4)12-10-6-2/h15-16H,5-14H2,1-4H3,(H,17,18)
Formula Weight 322.2273
Chemical Name or Material Bis(2-ethylhexyl) phosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.