missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-ethylhexyl) Phthalate (Phenyl-13C6, D4), TRC

Product Code. 30343822
Change view
Click to view available options
Quantity:
0.5 mg
5 mg
Unit Size:
0.5mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30343822 0.5 mg 0.5mg
30335504 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30343822 Supplier Toronto Research Chemicals Supplier No. TRCB4338510.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bis(2-ethylhexyl) Phthalate (Phenyl-13C6, D4)
Molecular Formula C1813C6H34D4O4
Quantity 0.5 mg
Synonym 1,2-Benzenedicarboxylic Acid 1,2-bis(2-ethylhexyl) Ester (Phenyl-13C6, D4), Bisoflex 81 (Phenyl-13C6, D4), Bisoflex DOP (Phenyl-13C6, D4), Di(2-ethylhexyl) Phthalate (Phenyl-13C6, D4), Di(isooctyl) Phthalate (Phenyl-13C6, D4), Etalon (Phenyl-13C6, D4), Octyl Phthalate (Phenyl-13C6, D4), Palatinol AH (Phenyl-13C6, D4), Phthalic Acid Dioctyl Ester(Phenyl-13C6, D4), DEHP (Phenyl-13C6, D4), Di(2-ethylhexyl) Phthalate (Phenyl-13C6, D4)
InChI Formula InChI=1S/C24H38O4/c1-5-9-13-19(7-3)17-27-23(25)21-15-11-12-16-22(21)24(26)28-18-20(8-4)14-10-6-2/h11-12,15-16,19-20H,5-10,13-14,17-18H2,1-4H3/i11+1D,12+1D,15+1D,16+1D,21+1,22+1
SMILES O=C([13C]1=[13C]([2H])[13C]([2H])=[13C]([2H])[13C]([2H])=[13C]1C(OCC(CC)CCCC)=O)OCC(CC)CCCC
IUPAC Name bis(2-ethylhexyl) 3,4,5,6-tetradeuterio(1,2,3,4,5,6-13C6)cyclohexa-2,4,6-triene-1,2-dicarboxylate
Molecular Weight (g/mol) 400.54
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.