missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-hydroxyethyl)dimethylammonium chloride, 99%

Product Code. 10219500 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10219500 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10219500 Supplier Thermo Scientific Acros Supplier No. 402530250
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 38402-02-7 | C6H16ClNO2 | 169.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Specifications

CAS 38402-02-7
CAS Min % 98.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Glass bottle
Molecular Formula C6H16ClNO2
Linear Formula (HOCH2CH2)2N(Cl)(CH3)2
MDL Number MFCD00041987
Quantity 25 g
Synonym bis 2-hydroxyethyl dimethylammonium chloride, 2-hydroxy-n-2-hydroxyethyl-n,n-dimethylethanaminium chloride, ethanaminium, 2-hydroxy-n-2-hydroxyethyl-n,n-dimethyl-, chloride, ethanaminium, 2-hydroxy-n-2-hydroxyethyl-n,n-dimethyl-, chloride 1:1, bis 2-hydroxyethyl dimethylazanium chloride, bis 2-hydroxyethyl-dimethylazanium chloride, n-2-hydroxyethyl-n,n-dimethyl-n-2-hydroxyethyl ammonium chloride
Solubility Information Solubility in water: soluble
InChI Key OPVXPOIGBSHXNM-UHFFFAOYSA-M
SMILES C[N+](C)(CCO)CCO.[Cl-]
IUPAC Name bis(2-hydroxyethyl)-dimethylazanium;chloride
Molecular Weight (g/mol) 169.65
PubChem CID 162303
Formula Weight 169.65
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Bis(2-hydroxyethyl)dimethylammonium chloride, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Bis(2-hydroxyethyl)dimethylammonium chloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.