missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-hydroxyethyl) disulfide, tech. 90%

Product Code. 11322627
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11322627 50 g 50g
11332627 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11322627 Supplier Thermo Scientific Alfa Aesar Supplier No. L11800.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1892-29-1 | C4H10O2S2 | 154.24 g/mol

Bis(2-hydroxyethyl) disulfide is used as an intermediate in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(2-hydroxyethyl) disulfide is used as an intermediate in chemical research.

Solubility
Soluble in water, ethanol, acetone, and ether.

Notes
Incompatible with strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1892-29-1
Molecular Formula C4H10O2S2
Molecular Weight (g/mol) 154.24
MDL Number MFCD00002906
InChI Key KYNFOMQIXZUKRK-UHFFFAOYSA-N
Synonym 2-hydroxyethyl disulfide, 2,2'-dithiodiethanol, bis 2-hydroxyethyl disulfide, ethanol, 2,2'-dithiobis, hydroxyethyl disulfide, diethanol disulfide, 2,2'-dithiobisethanol, 2-hydroxyethanedisulfide, usaf th-9, 2,2'-disulfanediyldiethanol
PubChem CID 15906
ChEBI CHEBI:43136
IUPAC Name 2-(2-hydroxyethyldisulfanyl)ethanol
SMILES OCCSSCCO

Specifications

Melting Point ∼25°C
Density 1.261
Boiling Point 158°C to 163°C (3.5 mmHg)
Flash Point >110°C (230°F)
Odor Unpleasant
Linear Formula HOCH2CH2S-SCH2CH2OH
Refractive Index 1.57
Quantity 50 g
UN Number UN2810
Beilstein 1735851
Solubility Information Soluble in water,ethanol,acetone,and ether.
Formula Weight 154.24
Percent Purity 90%
Grade Technical
Chemical Name or Material Bis(2-hydroxyethyl) disulfide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Bis(2-hydroxyethyl) disulfide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Bis(2-hydroxyethyl)disulfide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.