missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-methylallyl)(1,5-cyclooctadiene)ruthenium(II)

Product Code. 15477625
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity unitSize
15477625 1 g 1g
1 options
This item is not returnable. View return policy

Product Code. 15477625

Brand: Thermo Scientific Alfa Aesar H26891.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12289-94-0 | C16H26Ru | 319.45 g/mol

Bis(2-methylallyl)(1,5-cyclooctadiene)ruthenium(II) is used as a catalyst and pharmaceutical intermediate. It is also used as a precatalysts for asymmetric and Cross-Coupling Catalysis reactions, and in organic chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(2-methylallyl)(1,5-cyclooctadiene)ruthenium(II) is used as a catalyst and pharmaceutical intermediate. It is also used as a precatalysts for asymmetric and Cross-Coupling Catalysis reactions, and in organic chemical synthesis.
TRUSTED_SUSTAINABILITY

Specifications

CAS 12289-94-0
Melting Point 83°C to 86°C
Molecular Formula C16H26Ru
MDL Number MFCD00216965
Quantity 1 g
UN Number UN3395
Sensitivity Air and moisture sensitive
Synonym (1,5-Cyclooctadiene)bis(2-methylallyl)ruthenium(II); Ruthenium(II) bis(2-methylallyl) 1,5-cyclooctadiene complex
Solubility Information It is soluble in water.
InChI Key POYBJJLKGYXKJH-UHFFFAOYSA-N
SMILES [Ru++].Cc(:c):c.Cc(:c):c.C1C\C=C/CC\C=C/1
IUPAC Name (5Z)-cycloocta-1,5-diene;2-methanidylprop-1-ene;ruthenium(2+)
Molecular Weight (g/mol) 319.45
PubChem CID 91884701
Formula Weight 319.45
Chemical Name or Material Bis(2-methylallyl)(1,5-cyclooctadiene)ruthenium(II)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.