missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-oxo-3-oxazolidinyl)phosphinic chloride, 97%

Product Code. 11332706
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11332706 25 g 25g
11312706 1 g 1g
11322706 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11332706 Supplier Thermo Scientific Alfa Aesar Supplier No. L08775.14
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 68641-49-6 | C6H8ClN2O5P | 254.563 g/mol

Bis(2-oxo-3-oxazolidinyl)phosphinic chloride was used in the preparation of hexadepsipeptide. Also used as reagent is used for activating the carboxylic group, synthesis of amides, esters and peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(2-oxo-3-oxazolidinyl)phosphinic chloride was used in the preparation of hexadepsipeptide. Also used as reagent is used for activating the carboxylic group, synthesis of amides, esters and peptides.

Solubility
Hydrolyzes with water.

Notes
Moisture Sensitive, store away from moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68641-49-6
Molecular Formula C6H8ClN2O5P
Molecular Weight (g/mol) 254.563
MDL Number MFCD00010077
InChI Key KLDLRDSRCMJKGM-UHFFFAOYSA-N
Synonym bis 2-oxo-3-oxazolidinyl phosphinic chloride, bop-cl, bis 2-oxooxazolidin-3-yl phosphinic chloride, ccris 2607, phosphinic chloride, bis 2-oxo-3-oxazolidinyl, bis 2-oxo-3-oxazolidinyl phosphonic chloride, bis 2-oxo-1,3-oxazolidin-3-yl phosphinoyl chloride, bis-2-oxo-3-oxazolidinyl phosphinic chloride, n,n-bis 2-oxo-3-oxazolidinyl phosphinic chloride, bis 2-oxo-3-oxazolidinyl phosphinic chloride bop-cl
PubChem CID 152842
IUPAC Name 3-[chloro-(2-oxo-1,3-oxazolidin-3-yl)phosphoryl]-1,3-oxazolidin-2-one
SMILES C1COC(=O)N1P(=O)(N2CCOC2=O)Cl

Specifications

Melting Point ∽195°C (decomposition)
Quantity 25 g
UN Number UN3261
Beilstein 3654596
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes with water.
Formula Weight 254.57
Percent Purity 97%
Chemical Name or Material Bis(2-oxo-3-oxazolidinyl)phosphinic chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.