missing translation for 'onlineSavingsMsg'
Learn More

Bis[2-(perfluorohexyl)ethyl] phosphate, TRC

Product Code. 17962205
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17962205 10 mg 10mg
17991555 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17962205 Supplier Toronto Research Chemicals Supplier No. TRCB51630010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57677-95-9
Molecular Formula C16 H9 F26 O4 P
Molecular Weight (g/mol) 790.17
Synonym Bis[2-(perfluorohexyl)ethyl] Phosphate, 1-Octanol, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro-, 1,1'-(hydrogen phosphate) (ACI), 1-Octanol, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro-, hydrogen phosphate (9CI), 6:2 Polyfluoroalkylphosphoric acid diester, Bis(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro-1-octanol),hydrogen phosphate, Bis[2-(perfluorohexyl)ethyl] phosphate, 6:2 diPAP, Bis(1H,1H,2H,2H-perfluorooctyl)phosphate, 6:2 Di-PAPS
IUPAC Name bis(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl) hydrogen phosphate
SMILES OP(=O)(OCCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)OCCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C16H9F26O4P/c17-5(18,7(21,22)9(25,26)11(29,30)13(33,34)15(37,38)39)1-3-45-47(43,44)46-4-2-6(19,20)8(23,24)10(27,28)12(31,32)14(35,36)16(40,41)42/h1-4H2,(H,43,44)
Formula Weight 789.9823
Percent Purity >95%
Chemical Name or Material Bis[2-(perfluorohexyl)ethyl]phosphate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.