missing translation for 'onlineSavingsMsg'
Learn More

Bis[2-(succinimidooxycarbonyloxy)ethyl] Sulfone, TRC

Product Code. 30344428
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30344428 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30344428 Supplier Toronto Research Chemicals Supplier No. TRCB58500025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bis[2-(succinimidooxycarbonyloxy)ethyl] Sulfone
CAS 57683-72-4
Molecular Formula C14 H16 N2 O12 S
Quantity 25 mg
InChI Formula InChI=1S/C14H16N2O12S/c17-9-1-2-10(18)15(9)27-13(21)25-5-7-29(23,24)8-6-26-14(22)28-16-11(19)3-4-12(16)20/h1-8H2
SMILES O=C(OCCS(=O)(=O)CCOC(=O)ON1C(=O)CCC1=O)ON2C(=O)CCC2=O
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 2-[2-(2,5-dioxopyrrolidin-1-yl)oxycarbonyloxyethylsulfonyl]ethyl carbonate
Molecular Weight (g/mol) 436.35
Formula Weight 436.0424
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.