missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(3,5,3',5'-dimethoxydibenzylideneacetone)palladium(0), 97%
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
missing translation for 'unitSize'
1g
5g
Description
Bis(3,5,3',5'-dimethoxydibenzylideneacetone)palladium(0) used as catalyst in pharmaceutical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Bis(3,5,3′,5′-dimethoxydibenzylideneacetone)palladium(0) used as catalyst in pharmaceutical research.
Solubility
Insoluble in water.
Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
Bis(3,5,3′,5′-dimethoxydibenzylideneacetone)palladium(0) used as catalyst in pharmaceutical research.
Solubility
Insoluble in water.
Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
Specifications
Specifications
| Melting Point | 170°C to 178°C |
| Color | Red |
| Quantity | 5 g |
| Solubility Information | Insoluble in water. |
| IUPAC Name | 1,5-bis(3,5-dimethoxyphenyl)penta-1,4-dien-3-one;palladium |
| Formula Weight | 815.23 |
| Concentration or Composition (by Analyte or Components) | Pd 13% |
| Physical Form | Powder |
| Chemical Name or Material | Bis(3,5,3',5'-dimethoxydibenzylideneacetone)palladium(0) |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction