Learn More
Bis[3-(triethoxysilyl)propyl]tetrasulfide, S 22.3% (typical)
Description
Bis[3-(triethoxysilyl)propyl]tetrasulfide is used in radial tires, rubber tire treads, carcass, tire walls, solid rubber tires and other rubber products. It acts as a chemical intermediate and as a cross linking agent. Furthermore, it acts as a coupling agent used to chemically modify silica surface. In addition, it serves as a curing agent for good heat aging.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Density | 1.074 g/mL |
| Boiling Point | 250°C (decomposition) |
| Flash Point | 91°C (196°F) |
| Quantity | 25 g |
| Solubility Information | Miscible with toluene,primary alcohols,ketones,benzene,chlorinated hydrocarbons,acetonitrile,dimethylformaminde and dimethysulfoxide. Immiscible with water. |
| Formula Weight | 538.94 |
| Concentration or Composition (by Analyte or Components) | S 22.3% (typical) |
| Physical Form | Liquid |
| Chemical Name or Material | Bis[3- (triethoxysilyl)propyl]tetrasulfide |
Safety and Handling
- Bis(3-(triethoxysilyl)propyl)tetrasulfide
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contain: Bis(3-(triethoxysilyl)propyl)tetrasulfide
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.