missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-methoxyphenyl)disulfide, 97%

Product Code. 11497076
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11497076 5 g 5g
11487076 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11497076 Supplier Thermo Scientific Alfa Aesar Supplier No. B25399.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5335-87-5 | C14H14O2S2 | 278.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 5335-87-5
Molecular Formula C14H14O2S2
Molecular Weight (g/mol) 278.38
MDL Number MFCD00041358
InChI Key PZQGLCGLPMWYBT-UHFFFAOYSA-N
Synonym bis 4-methoxyphenyl disulfide, disulfide, bis 4-methoxyphenyl, p-methoxyphenyl disulfide, 4,4'-dimethoxy diphenyldisulfide, 4-methoxyphenyl disulfide, anisole, 4,4'-dithiodi, 1-methoxy-4-4-methoxyphenyl disulfanyl benzene, 1,2-bis 4-methoxyphenyl disulfane, bis p-methoxyphenyl disulfide
PubChem CID 79258
IUPAC Name 1-methoxy-4-[(4-methoxyphenyl)disulfanyl]benzene
SMILES COC1=CC=C(SSC2=CC=C(OC)C=C2)C=C1

Especificaciones

Melting Point 37°C to 41°C
Boiling Point 200°C (3 mmHg)
Quantity 5 g
Beilstein 1914789
Formula Weight 278.39
Percent Purity 97%
Chemical Name or Material Bis(4-methoxyphenyl)disulfide

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.