missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-nitrophenyl) carbonate, 97%

Product Code. 10362160 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 10362160

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5070-13-3 | C13H8N2O7 | 304.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5070-13-3
Molecular Formula C13H8N2O7
Molecular Weight (g/mol) 304.21
MDL Number MFCD00007322
InChI Key ACBQROXDOHKANW-UHFFFAOYSA-N
Synonym bis 4-nitrophenyl carbonate, bis p-nitrophenyl carbonate, 4,4'-dinitrodiphenyl carbonate, carbonic acid, bis 4-nitrophenyl ester, p,p'-dinitrodiphenylcarbonate, carbonic acid bis 4-nitrophenyl ester, carbonic acid, bis p-nitrophenyl ester, npc bis 4-nitrophenyl carbonate
PubChem CID 78756
IUPAC Name bis(4-nitrophenyl) carbonate
SMILES C1=CC(=CC=C1[N+](=O)[O-])OC(=O)OC2=CC=C(C=C2)[N+](=O)[O-]

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 139°C to 141°C
Color Brown to Green or Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (O2NC6H4O)2CO
Quantity 1 g
Beilstein 06, I, 120
Fieser 01,330
Formula Weight 304.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Bis(4-nitrophenyl) carbonate
Show More Show Less
compliance-icons
Product Identifier
  • Bis(4-nitrophenyl) carbonate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.