missing translation for 'onlineSavingsMsg'
Learn More

Bis(acetonitrile)dichloropalladium(II), Pd 40.5%

Product Code. 11396697
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11396697 5 g 5g
11306707 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11396697 Supplier Thermo Scientific Alfa Aesar Supplier No. 010002.06
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Olefin isomerization, preparation of acetals and hemiacetal esters | CAS: 14592-56-4 | C4H6Cl2N2Pd | 259.42 g/mol

It is used as a catalyst in an ether-directed aza-Claisen rearrangement. Also used in cyclization and cross-coupling reactions. It is applied for olefin isomerization, preparation of acetals and hemiacetal esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14592-56-4
Molecular Formula C4H6Cl2N2Pd
Molecular Weight (g/mol) 259.42
MDL Number MFCD00013122
InChI Key XGPJSRXOBCGBMN-UHFFFAOYSA-L
Synonym cis-Dichlorobis(acetonitrile)palladium(II)
IUPAC Name palladium(2+) bis(acetonitrile) dichloride
SMILES CC#N.CC#N.Cl[Pd++]Cl

Specifications

Odor Odorless
Quantity 5 g
UN Number UN3439
Solubility Information Soluble in acetonitrile, acetone, chloroform
Formula Weight 259.41
Concentration or Composition (by Analyte or Components) Pd 40.5%
Physical Form Powder
Chemical Name or Material Bis(acetonitrile)dichloropalladium(II)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.