missing translation for 'onlineSavingsMsg'
Learn More

Melford bis-Acrylamide

Product Code. p-200087230
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30017948 25 g 25g
30017947 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30017948 Supplier Melford Supplier No. A1127025.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.

  • CAS Number: 110-26-9
  • Molecular Formula: C7H10N2O2
  • Molecular Weight: 154.2

Specifications

Chemical Name or Material bis-Acrylamide
CAS 110-26-9
Quantity 25 g
Molecular Formula C7H10N2O2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.