missing translation for 'onlineSavingsMsg'
Learn More

Bis-acrylamide (Electrophoresis), Fisher BioReagents

Product Code. 10689923
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Poly Bottle
Unit Size:
100g
25g
Product Code. Quantity Packaging unitSize
10689923 25 g Poly Bottle 25g
10689733 100 g Poly Bottle 100g
2 options
This item is not returnable. View return policy

Product Code. 10689923

Brand: Fisher BioReagents BP17125

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
  • Bis-acrylamide is a cross-linking agent used in the preparation of polyacrylamide gels for electrophoresis
  • RNase, DNase, and protease free
  • Specified and tested for use in electrophoresis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-26-9
Molecular Formula C7H10N2O2
Molecular Weight (g/mol) 154.169
InChI Key ZIUHHBKFKCYYJD-UHFFFAOYSA-N
Synonym N, N«–Methylene-bis-acrylamide
PubChem CID 8041
IUPAC Name N-[(prop-2-enoylamino)methyl]prop-2-enamide
SMILES C=CC(=O)NCNC(=O)C=C

Specifications

Absorbance 0.2 AU max., 0.2 max. (1% solution) at 290nm
Physical Form Solid
Melting Point 300°C
Color White
pH 5
Quantity 25 g
DNase DNase free
Packaging Poly Bottle
Assay Percent Range ≥95 %
Linear Formula CH2CHCONHCH2NHCOCHCH2
Formula Weight 154.17
Percent Purity ≥95%
Grade Electrophoresis
Chemical Name or Material Bis-Acrylamide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.