missing translation for 'onlineSavingsMsg'
Learn More

Bis-acrylamide Solution (2% w/v/Electrophoresis), Fisher BioReagents

Product Code. 10193523
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10193523 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10193523 Supplier Fisher BioReagents Supplier No. BP1404250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Bis-Acrylamide Solution is a cross-linker commonly used in the preparation of polyacrylamide electrophoresis gels.
TRUSTED_SUSTAINABILITY

Specifications

Physical Form Liquid
CAS 110-26-9, 7732-18-5
Color Colorless
Quantity 250 mL
Packaging Amber Glass
Molecular Formula C7H10N2O2
InChI Key ZIUHHBKFKCYYJD-UHFFFAOYSA-N
SMILES C=CC(=O)NCNC(=O)C=C
IUPAC Name N-[(prop-2-enoylamino)methyl]prop-2-enamide
Molecular Weight (g/mol) 154.169
PubChem CID 8041
Formula Weight 154.08
Grade Electrophoresis
Chemical Name or Material Bis-Acrylamide Solution, 2% w/v
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.