missing translation for 'onlineSavingsMsg'
Learn More

bis-Benzimide H-33342 trihydrochloride trihydrate, 98%

Product Code. 15490227
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15490227 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15490227 Supplier Thermo Scientific Alfa Aesar Supplier No. J62134.MC
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 23491-52-3 | C27H28N6O | 452.562 g/mol

Stain for identifying specific chromosome banding patterns

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 23491-52-3
Color Yellow to Green
Molecular Formula C27H28N6O
MDL Number MFCD00012679
Quantity 100 mg
Beilstein 4088183
Synonym hoechst 33342, bisbenzimide, hoechst33342, unii-99kzs6cnzx, 99kzs6cnzx, 2'-4-ethoxyphenyl-5-4-methylpiperazin-1-yl-1h,1'h-2,5'-bibenzo d imidazole, 2'-4-ethoxyphenyl-5-4-methyl-1-piperazinyl-2,5'-bi-benzimidazole, 2'-4-ethoxyphenyl-5-4-methyl-1-piperazinyl-2,5'-bi-1h-benzimidazole, 2'-4'-ethoxyphenyl-5-4-methylpiperazin-1-yl-2,5'-bis-1h-benzimidazole trihydrochloride trihydrate, 2'-p-ethoxyphenyl-5-4-methyl-1-piperazinyl-2,5'-bibenzimidazole
InChI Key PRDFBSVERLRRMY-UHFFFAOYSA-N
SMILES CCOC1=CC=C(C=C1)C2=NC3=C(N2)C=C(C=C3)C4=NC5=C(N4)C=C(C=C5)N6CCN(CC6)C
IUPAC Name 2-(4-ethoxyphenyl)-6-[6-(4-methylpiperazin-1-yl)-1H-benzimidazol-2-yl]-1H-benzimidazole
Molecular Weight (g/mol) 452.562
PubChem CID 1464
ChEBI CHEBI:51232
Formula Weight 615.99 (561.94 Anhydrous)
Percent Purity 98%
Physical Form Powder
Chemical Name or Material bis-Benzimide H-33342 trihydrochloride trihydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.