missing translation for 'onlineSavingsMsg'
Learn More

Bis(benzonitrile)palladium(II) chloride, 97+%

Product Code. 10667874 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
250mg
This item is not returnable. View return policy

Product Code. 10667874

Brand: Thermo Scientific Acros 207900100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14220-64-5 | C14H10Cl2N2Pd | 383.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14220-64-5
Molecular Formula C14H10Cl2N2Pd
Molecular Weight (g/mol) 383.57
MDL Number MFCD00013123
InChI Key QQPSWGUJSPTOSH-UHFFFAOYSA-L
Synonym bis benzonitrile palladium chloride, bis benzonitrile palladium ii chloride, trans-bis benzonitrile palladium ii chloride, bis benzonitrile palladium ii chloride, 500mg, palladium 2+ bis benzonitrile dichloride
PubChem CID 10271322
IUPAC Name benzonitrile;palladium(2+);dichloride
SMILES Cl[Pd++](Cl)([N]#CC1=CC=CC=C1)[N]#CC1=CC=CC=C1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 120.0°C
Color Orange to Brown
Elemental Analysis 99.0% min.
Infrared Spectrum Authentic
Assay Percent Range 97+%
Packaging Glass bottle
Linear Formula (C6H5CN)2PdCl2
Quantity 10 g
Fieser 01,56; 05,31; 06,45; 09,44; 10,31; 13,34
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acetone, benzene, toluene, chloroform
Formula Weight 383.55
Percent Purity 97+%
Physical Form Crystalline Powder
Chemical Name or Material Bis(benzonitrile)palladium(II) chloride
Show More Show Less
compliance-icons
Product Identifier
  • Bis(benzonitrile)palladium(II) chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H413-May cause long lasting harmful effects to aquatic life.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.