missing translation for 'onlineSavingsMsg'
Learn More

Bis(butoxyethyl) Phosphate, TRC

Product Code. 30003602
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003602 100 mg 100mg
30007352 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30003602 Supplier Toronto Research Chemicals Supplier No. TRCB415125100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14260-97-0
Molecular Formula C12H27O6P
Molecular Weight (g/mol) 298.31
Synonym Ethanol, 2-butoxy-, hydrogen phosphate (7CI, 8CI, 9CI), Bis(2-butoxyethyl) phosphate, Bis(butoxyethyl) phosphate, Bis(2-butoxyethyl)hydrogenphosphate, Bis(2-butoxyethyl) Phosphate, 2-Butoxy-ethanol Hydrogen Phosphate, Bis(2-butoxyethyl) Phosphate, 2-Butoxy-ethanol 1,1'-(Hydrogen Phosphate)
IUPAC Name bis(2-butoxyethyl) hydrogen phosphate
SMILES CCCCOCCOP(=O)(O)OCCOCCCC

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C12H27O6P/c1-3-5-7-15-9-11-17-19(13,14)18-12-10-16-8-6-4-2/h3-12H2,1-2H3,(H,13,14)
Formula Weight 298.1545
Chemical Name or Material Bis(butoxyethyl) Phosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.